EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
HAX-1 regulates cyclophilin-D levels and mitochondria permeability transition pore in the heart.
PMID 26553996
Abstract
The major underpinning of massive cell death associated with myocardial infarction involves opening of the mitochondrial permeability transition pore (mPTP), resulting in disruption of mitochondria membrane integrity and programmed necrosis. Studies in human lymphocytes suggested that the hematopoietic-substrate-1 associated protein X-1 (HAX-1) is linked to regulation of mitochondrial membrane function, but its role in controlling mPTP activity remains obscure. Herein we used models with altered HAX-1 expression levels in the heart and uncovered an unexpected role of HAX-1 in regulation of mPTP and cardiomyocyte survival. Cardiac-specific HAX-1 overexpression was associated with resistance against loss of mitochondrial membrane potential, induced by oxidative stress, whereas HAX-1 heterozygous deficiency exacerbated vulnerability. The protective effects of HAX-1 were attributed to specific down-regulation of cyclophilin-D levels leading to reduction in mPTP activation. Accordingly, cyclophilin-D and mPTP were increased in heterozygous hearts, but genetic ablation of cyclophilin-D in these hearts significantly alleviated their susceptibility to ischemia/reperfusion injury. Mechanistically, alterations in cyclophilin-D levels by HAX-1 were contributed by the ubiquitin-proteosomal degradation pathway. HAX-1 overexpression enhanced cyclophilin-D ubiquitination, whereas proteosomal inhibition restored cyclophilin-D levels. The regulatory effects of HAX-1 were mediated through interference of cyclophilin-D binding to heat shock protein-90 (Hsp90) in mitochondria, rendering it susceptible to degradation. Accordingly, enhanced Hsp90 expression in HAX-1 overexpressing cardiomyocytes increased cyclophilin-D levels, as well as mPTP activation upon oxidative stress. Taken together, our findings reveal the role of HAX-1 in regulating cyclophilin-D levels via an Hsp90-dependent mechanism, resulting in protection against activation of mPTP and subsequent cell death responses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
860336
3,3′,5,5′-Tetramethylbenzidine, ≥99%
C16H20N2
T2885
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
C16H20N2
T5525
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
C16H20N2
87748
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
C16H20N2
A0781
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
C20H34O4
C0780
Creatine, anhydrous
C4H9N3O2
C3662
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
C62H111N11O12
C1832
Cyclosporin A, from Tolypocladium inflatum, BioReagent, for molecular biology, ≥95%
C62H111N11O12
30024
Cyclosporin A, ≥98.5% (HPLC)
C62H111N11O12
316989
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
H2O2
H1009
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
H2O2
40287
Hydrogen peroxide solution, semiconductor grade MOS PURANAL (Honeywell 17937), ≥30%
H2O2
18304
Hydrogen peroxide solution, meets analytical specification of Ph. Nord., 34.5-36.5%
H2O2
C0158
L-Carnitine inner salt, synthetic, ≥98%
C7H15NO3
G3126
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
C5H10N2O3
G5792
L-Glutamine, PharmaGrade, Ajinomoto, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C5H10N2O3
G8540
L-Glutamine, meets USP testing specifications, cell culture tested, 99.0-101.0%, from non-animal source
C5H10N2O3
G6392
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
C5H10N2O3
49419
L-Glutamine, BioUltra, ≥99.5% (NT)
C5H10N2O3
W368401
L-Glutamine
C5H10N2O3
295574
Nitrogen, ≥99.998%
N2
P7626
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
C7H7FO2S
78830
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
C7H7FO2S
P2222
Potassium phosphate dibasic, meets USP testing specifications
HK2O4P
450200
Potassium phosphate dibasic, 99.95% trace metals basis
HK2O4P
P3786
Potassium phosphate dibasic, ACS reagent, ≥98%
HK2O4P
P8281
Potassium phosphate dibasic, reagent grade, ≥98.0%
HK2O4P
60353
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
HK2O4P
60356
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
HK2O4P
P8584
Potassium phosphate dibasic solution, 1.0 M
HK2O4P