EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Transmembrane Topologies of Ca2+-permeable Mechanosensitive Channels MCA1 and MCA2 in Arabidopsis thaliana.
PMID 26555262
Abstract
Sensing mechanical stresses, including touch, stretch, compression, and gravity, is crucial for growth and development in plants. A good mechanosensor candidate is the Ca(2+)-permeable mechanosensitive (MS) channel, the pore of which opens to permeate Ca(2+) in response to mechanical stresses. However, the structure-function relationships of plant MS channels are poorly understood. Arabidopsis MCA1 and MCA2 form a homotetramer and exhibit Ca(2+)-permeable MS channel activity; however, their structures have only been partially elucidated. The transmembrane topologies of these ion channels need to be determined in more detail to elucidate the underlying regulatory mechanisms. We herein determined the topologies of MCA1 and MCA2 using two independent methods, the Suc2C reporter and split-ubiquitin yeast two-hybrid methods, and found that both proteins are single-pass type I integral membrane proteins with extracellular N termini and intracellular C termini. These results imply that an EF hand-like motif, coiled-coil motif, and plac8 motif are all present in the cytoplasm. Thus, the activities of both channels can be regulated by intracellular Ca(2+) and protein interactions.