EMAIL THIS PAGE TO A FRIEND
AMB Express
High-level fed-batch fermentative expression of an engineered Staphylococcal protein A based ligand in E. coli: purification and characterization.
PMID 26556030
Abstract
The major platform for high level recombinant protein production is based on genetically modified microorganisms like Escherichia coli (E. coli) due to its short dividing time, ability to use inexpensive substrates and additionally, its genetics is comparatively simple, well characterized and can be manipulated easily. Here, we investigated the possibilities of finding the best media for high cell density fermentation, by analyzing different media samples, focusing on improving fermentation techniques and recombinant protein production. Initial fermentation of E. coli BL21 DE3:pAV01 in baffled flasks showed that high cell density was achieved when using complex media, Luria-Bertani (LB) and Terrific medium broth (TB) (10 and 14xa0g/L wet weight, respectively), as compared to mineral media M9, modified minimal medium (MMM) and Riesenberg mineral medium (RM) (7, 8 and 7xa0g/L, respectively). However, in fed-batch fermentation processes when using MMM after 25xa0h cultivation, it was possible to yield an optical density (OD600) of 139 corresponding to 172xa0g/L of wet biomass was produced in a 30xa0L TV Techfors-S Infors HT fermenter, with a computer controlled nutrient supply (glucose as a carbon source) delivery system, indicating nearly 1.5 times that obtained from TB. Upon purification, a total of 1.65xa0mg/g of protein per gram cell biomass was obtained and the purified AviPure showed affinity for immunoglobulin. High cell density fed batch fermentation was achieved by selecting the best media and growth conditions, by utilizing a number of fermentation parameters like media, fermentation conditions, chemical concentrations, pO2 level, stirrer speed, pH level and feed media addition. It is possible to reach cell densities higher than shake flasks and stirred tank reactors with the improved oxygen transfer rate and feed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0918 Terrific Broth, modified, powder