EMAIL THIS PAGE TO A FRIEND
Scientific Reports
Quercitrin for periodontal regeneration: effects on human gingival fibroblasts and mesenchymal stem cells.
PMID 26558438
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H1758
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
HCl
435570
Hydrochloric acid, reagent grade, 37%
HCl
40233
Hydrochloric acid, semiconductor grade PURANAL (Honeywell 17863), ≥32%
HCl
40253
Hydrochloric acid, semiconductor grade PURANAL (Honeywell 17823), fuming 37%, 37-38%
HCl
84429
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
HCl
H9892
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
HCl
84434
Hydrochloric acid solution, BioXtra, ~0.1 M in H2O
HCl
H0636
Hydrochloric acid solution, protein sequencing grade, liquid
HCl
2104
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
HCl
W530574
Hydrochloric acid solution, 32 wt. % in H2O, FCC
HCl
295426
Hydrogen chloride, ReagentPlus®, ≥99%
HCl
705551
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
HCl
A5960
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
C6H8O6
A7506
L-Ascorbic acid, reagent grade, crystalline
C6H8O6
A0278
L-Ascorbic acid, reagent grade
C6H8O6
A4403
L-Ascorbic acid, powder, cell culture tested, γ-irradiated
C6H8O6
A4544
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
C6H8O6
A2218
L-Ascorbic acid, meets USP testing specifications
C6H8O6
A2174
L-Ascorbic acid, plant cell culture tested
C6H8O6
A2343
L-Ascorbic acid, meets USP testing specifications
C6H8O6
95209
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
C6H8O6
33034
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.7-100.5% (oxidimetric)
C6H8O6
95210
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
C6H8O6
47863
L-Ascorbic acid, analytical standard
C6H8O6
255564
L-Ascorbic acid, ACS reagent, ≥99%
C6H8O6
A92902
L-Ascorbic acid, 99%
C6H8O6
W210901
L-Ascorbic acid, FCC, FG
C6H8O6
40104-U Methanolic HCl, 3 M HCl in methanol (3N), for GC derivatization, pkg of 1 × 2 L
HCl
P5640
Prostaglandin E2, ≥93% (HPLC), synthetic
C20H32O5
P6532
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
C20H32O5