EMAIL THIS PAGE TO A FRIEND
Soft matter
Toward the dynamic phase transition mechanism of a thermoresponsive ionic liquid in the presence of different thermoresponsive polymers.
PMID 26558815
Abstract
The influence of two thermoresponsive polymers, poly(N-isopropylacrylamide) (PNIPAM) and poly(N-vinylcaprolactam) (PVCL), on the phase transition behavior of a thermoresponsive ionic liquid, tributylhexylphosphonium 3-sulfopropylmethacrylate ([P4,4,4,6][MC3S]), was investigated. An obvious distinction was observed in the LCSTs and morphologies of [P4,4,4,6][MC3S]-PNIPAM and [P4,4,4,6][MC3S]-PVCL aqueous solutions, indicating their large differences in dynamic transition processes. In general, PNIPAM can "break" the water structure of [P4,4,4,6][MC3S] to decrease the transition temperature, while PVCL can "make" the water structure to increase it. Surprisingly, [P4,4,4,6][MC3S] has an unusual over-hydration behavior before dehydration while PNIPAM experiences a two-step transition process in [P4,4,4,6][MC3S]-PNIPAM aqueous solution, which has never been reported so far. Further studies revealed that the formation of strong intra-/inter-molecular hydrogen bonds C[double bond, length as m-dash]OD-N in PNIPAM is the driving force for the LCST phenomenon of [P4,4,4,6][MC3S]-PNIPAM solution, while it is the [P4,4,4,6][MC3S] that dominates the phase separation of [P4,4,4,6][MC3S]-PVCL solution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
415464
N-Vinylcaprolactam, 98%
C8H13NO