EMAIL THIS PAGE TO A FRIEND
Scientific Reports
Label-free colorimetric detection of mercury via Hg(2+) ions-accelerated structural transformation of nanoscale metal-oxo clusters.
PMID 26559602
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282979
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
C7H12O2
379115
DCC, 1.0 M in methylene chloride
C13H22N2
D80002
DCC, 99%
C13H22N2
633267
Dicyclohexylcarbodiimide solution, 60 wt. % in xylenes
C13H22N2
M1770
Methanol, BioReagent, suitable for protein sequencing
CH4O
19-2470
Methanol, for HPLC
CH4O
19-2453
Methanol, for residue analysis, JIS 5000
CH4O
19-2400
Methanol, SAJ special grade
CH4O
19-2425
Methanol, suitable for determination of dioxins
CH4O
717975
Methanol, NMR reference standard
CH4O
650188
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
CH4O
632546
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, for mass spectrometry, ≥99.5%
CH4O
667390
Methanol solution, contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) water, for HPLC
CH4O
721964
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
CH4O
721956
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
CH4O
606723
Methanol-12C, 99.95 atom % 12C
12CH4O
71687
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
HNaO
757527
Sodium hydroxide, ultra dry, powder, 99.99% trace metals basis
HNaO
72068
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
HNaO
S2567
Sodium hydroxide solution, volumetric, 1.0 M NaOH (1.0N)
HNaO
S2770
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
HNaO
609765
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
HNa16O