EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Porous lead(II)-based metal organic nanotubes as an adsorbent for dispersive solid-phase extraction of polybrominated diphenyl ethers from environmental water samples.
PMID 26559618
Abstract
Porous lead(II)-based metal-organic nanotubes (Pb-MONTs) were, for the first time, used as an adsorbent for dispersive solid-phase extraction (d-SPE) of polybrominated diphenyl ethers (PBDEs) at trace levels from environmental water samples. Gas chromatography-negative chemical ionization-mass spectrometry (GC-NCI-MS) was applied for sample detection. Box-Behnken design was performed to investigate and optimize the d-SPE parameters through a response surface methodology. The optimized conditions were obtained as listed: extraction time of 1min, 51.4mg of adsorbent and NaCl concentration of 7.42%. Under the optimized conditions, the new d-SPE-GC-NCI-MS method achieved wide range of linearity (2-1000ngL(-1)), low limits of detection (0.08-0.22ngL(-1)), satisfactory repeatability (0.79-8.62%, n=6) and satisfactory reproducibility (3.18-11.0%, n=5), and the possible extraction mechanism was also discussed. The proposed method was used in the analysis of real environmental water samples, and satisfactory recoveries were obtained in the range of 80.5-119.2%. These results indicated that Pb-MONTs have great potential as an adsorbent for the d-SPE of PBDEs at trace levels from environmental water samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
105899
Cyclohexanol, ReagentPlus®, 99%
C6H12O
57180
Indo 1, for fluorescence, ≥80.0% (HPLC)
C32H26K5N3O12