PLoS ONE
Seasonal Dynamics in the Chemistry and Structure of the Fat Bodies of Bumblebee Queens.
PMID 26559946
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1378
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
C15H24O
I9516
2-Propanol, BioReagent, for molecular biology, ≥99.5%
C3H8O
278475
2-Propanol, anhydrous, 99.5%
C3H8O
675431
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
C3H8O
437522
2-Propanol, ACS reagent, ≥99.5%
C3H8O
59304
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
C3H8O
40301
2-Propanol, semiconductor grade VLSI PURANAL (Honeywell 17604)
C3H8O
40219
2-Propanol, semiconductor grade PURANAL (Honeywell 17829)
C3H8O
270490
2-Propanol, 99.5%, HPLC grade
C3H8O
15-2360
2-Propanol, for HPLC
C3H8O
733458
2-Propanol, electronic grade, 99.999% trace metals basis
C3H8O
A1542
Ammonium acetate, for molecular biology, ≥98%
C2H7NO2
A7330
Ammonium acetate, BioXtra, ≥98%
C2H7NO2
372331
Ammonium acetate, 99.999% trace metals basis
C2H7NO2
431311
Ammonium acetate, ≥99.99% trace metals basis
C2H7NO2
238074
Ammonium acetate, ACS reagent, ≥97%
C2H7NO2
A7262
Ammonium acetate, reagent grade, ≥98%
C2H7NO2
A2706
Ammonium acetate solution, for molecular biology, 7.5 M
C2H7NO2
W218405
Butylated hydroxytoluene, ≥99%, FCC, FG
C15H24O
C7559
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
CHCl3
372978
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
CHCl3
288306
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
439142
Chloroform, for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
437581
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
480150
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
CHCl3
270636
Chloroform, for HPLC, ≥99.8%
CHCl3
48520-U
Chloroform, analytical standard
CHCl3
05-3433
Chloroform, for residue analysis, JIS 5000
CHCl3
05-3450
Chloroform, for HPLC
CHCl3
296082
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
C4H10O