EMAIL THIS PAGE TO A FRIEND
Bioresource Technology
Effects of cytoplasm and reactant polarities on acid-catalyzed lipid transesterification in wet microalgal cells subjected to microwave irradiation.
PMID 26562690