EMAIL THIS PAGE TO A FRIEND
Journal of clinical immunology
Early Complement Component Deficiency in a Single-Centre Cohort of Pediatric Onset Lupus.
PMID 26563161
Abstract
To assess complement levels C1q, C2, C3 and C4 in children with pediatric-onset lupus during the quiescent stage of disease. Thirty-four consecutive children with pediatric-onset SLE (onset below 12xa0years), in the quiescent stage were enrolled for the study. Twenty-nine age and sex matched healthy children were also enrolled for the purpose of comparison. Complement C1q and C2 levels were estimated by enzyme-linked immunosorbent assay (ELISA) whereas C3 and C4 were measured by end-point nephelometry. Genetic mutation analysis and functional assessment of classical complement pathway by ELISA were carried out in children with depressed levels of these complements. The study protocol was approved by the Institute Thesis Committee and the Institute Ethics Committee. Mean complement C1q, C2, C3 and C4 levels were 50.32, 17.28, 1320 and 236xa0mg/L respectively. Levels of complements were low in 7/34 children with SLE. An early age at onset, low anti-dsDNA titres and predominant skin manifestations were noted in children with decreased levels of complement C1q. Mutation analysis of C1qA gene revealed a homozygous nonsense mutation: C1QA (NM_015991) c.622C>T, p.Q208X in one child. A homozygous acceptor splice site mutation at the -2 position of intron2 of C1QA (c.164-2A>C) was detected in another child. The age at onset of disease was early in both these children, at 2.5 and 1.5xa0years respectively. Children with inherited deficiency of C1q and other early complement components present with early onset lupus that has a distinct clinical and immunological profile.