EMAIL THIS PAGE TO A FRIEND
Scientific reports
Cardiac Optogenetics: Enhancement by All-trans-Retinal.
PMID 26568132
Abstract
All-trans-Retinal (ATR) is a photosensitizer, serving as the chromophore for depolarizing and hyperpolarizing light-sensitive ion channels and pumps (opsins), recently employed as fast optical actuators. In mammalian optogenetic applications (in brain and heart), endogenous ATR availability is not considered a limiting factor, yet it is unclear how ATR modulation may affect the response to optical stimulation. We hypothesized that exogenous ATR may improve light responsiveness of cardiac cells modified by Channelrhodopsin2 (ChR2), hence lowering the optical pacing energy. In virally-transduced (Ad-ChR2(H134R)-eYFP) light-sensitive cardiac syncytium in vitro, ATR supplements ≤2 μM improved cardiomyocyte viability and augmented ChR2 membrane expression several-fold, while >4 μM was toxic. Employing integrated optical actuation (470 nm) and optical mapping, we found that 1-2 μM ATR dramatically reduced optical pacing energy (over 30 times) to several μW/mm(2), lowest values reported to date, but also caused action potential prolongation, minor changes in calcium transients and no change in conduction. Theoretical analysis helped explain ATR-caused reduction of optical excitation threshold in cardiomyocytes. We conclude that cardiomyocytes operate at non-saturating retinal levels, and carefully-dosed exogenous ATR can enhance the performance of ChR2 in cardiac cells and yield energy benefits over orders of magnitude for optogenetic stimulation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74875
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
C24H21Na3O11S3
R2500
all trans-Retinal, powder, ≥98%
C20H28O
C3607
Cyanocobalamin, meets USP testing specifications
C63H88CoN14O14P
47869
Cyanocobalamin (B12), analytical standard
C63H88CoN14O14P
47829
D-(+) Glucose, analytical standard
C6H12O6
G8270
D-(+)-Glucose, ≥99.5% (GC)
C6H12O6
G7021
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
C6H12O6
G7528
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
C6H12O6
G5767
D-(+)-Glucose, ACS reagent
C6H12O6
49139
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
C6H12O6
G6152
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
C6H12O6
G5146
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
C6H12O6
47249
D-(+)-Glucose, analytical standard
C6H12O6
D9434
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
C6H12O6
D8418
Dimethyl sulfoxide, for molecular biology
C2H6OS
D2438
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
C2H6OS
D4540
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
C2H6OS
D1435
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
C2H6OS
41639
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
C2H6OS
D9170
Dimethyl sulfoxide, PCR Reagent
C2H6OS
673439 Dimethyl sulfoxide solution, 50 wt. % in H2O
C2H6OS
H3375
HEPES, ≥99.5% (titration)
C8H18N2O4S
H4034
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
C8H18N2O4S
PHG0001
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O4S
RES6003H-B7
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O4S
H7523
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
C8H18N2O4S
54457
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
C8H18N2O4S
H6147
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
C8H18N2O4S
RDD002
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
C8H18N2O4S
83264
HEPES buffer solution, 1 M in H2O