EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Gd(III) complexes intercalated into hydroxy double salts as potential MRI contrast agents.
PMID 26568157
Abstract
The ion exchange intercalation of two Gd-based magnetic resonance imaging contrast agents into hydroxy double salts (HDSs) is reported. The presence of Gd(3+) diethylenetriaminepentaacetate and Gd(3+) diethylenetriaminepenta(methylenephosphonate) complexes in the HDS lattice after intercalation was confirmed by microwave plasma-atomic emission spectroscopy. The structural aspects of the HDS-Gd composites were studied by X-ray diffraction, with the intercalates having an interlayer spacing of 14.5-18.6 Å. Infrared spectroscopy confirmed the presence of characteristic vibration peaks associated with the Gd(3+) complexes in the intercalation compounds. The proton relaxivities of the Gd(3+) complex-loaded composites were 2 to 5-fold higher in longitudinal relaxivity, and up to 10-fold higher in transverse relaxivity, compared to solutions of the pure complexes. These data demonstrate that the new composites reported here are potentially potent MRI contrast agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282979
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
C7H12O2
D6518
Diethylenetriaminepentaacetic acid, ≥99% (titration)
C14H23N3O10
D1133
Diethylenetriaminepentaacetic acid, ≥98% (titration)
C14H23N3O10
263087
Gadolinium, chips
Gd
263060
Gadolinium, −40 mesh, 99% trace rare earth metals basis
Gd
GF55248453
Gadolinium, rod, 100mm, diameter 12.5mm, cast, 99%
Gd
GF88601531
Gadolinium, rod, 100mm, diameter 3.0mm, cast, 99%
Gd
GF96799875
Gadolinium, rod, 100mm, diameter 6.35mm, cast, 99%
Gd
GF99984335
Gadolinium, rod, 25mm, diameter 12.5mm, cast, 99%
Gd
GF48337487
Gadolinium, rod, 48mm, diameter 3.0mm, cast, 99%
Gd
GF98041989
Gadolinium, rod, 50mm, diameter 12.5mm, cast, 99%
Gd
GF15174390
Gadolinium, rod, 50mm, diameter 6.35mm, cast, 99%
Gd
GF82068790
Gadolinium, wire reel, 100mm, diameter 0.5mm, hard, 99.9%
Gd
GF13286369
Gadolinium, wire reel, 25mm, diameter 0.5mm, hard, 99.9%
Gd
GF52231711
Gadolinium, wire reel, 50mm, diameter 0.5mm, hard, 99.9%
Gd
693723
Gadolinium, foil, thickness 1 mm, 99.9% trace rare earth metals basis
Gd
261114
Gadolinium, ingot, 99.9% trace rare earth metals basis
Gd
263109
Lanthanum, powder, −40 mesh, under oil, 99.9% trace rare earth metals basis
La
263117
Lanthanum, pieces
La
261130
Lanthanum, ingot (under oil), 99.9% trace rare earth metals basis
La
GF13609111
Lanthanum, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
La
GF60507019
Lanthanum, rod, 50mm, diameter 6.35mm, cast, 99%
La
GF43011391
Lanthanum, foil, 100x100mm, thickness 0.125mm, as rolled, 99%
La
GF15415691
Lanthanum, foil, 100x100mm, thickness 0.1mm, as rolled, 99%
La
GF09165786
Lanthanum, foil, 25x25mm, thickness 0.1mm, as rolled, 99%
La
GF88400189
Lanthanum, foil, 29x33mm, thickness 0.5mm, as rolled, 99%
La
GF16232295
Lanthanum, foil, 35x47mm, thickness 0.5mm, as rolled, 99%
La
GF76465176
Lanthanum, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
La
GF45723326
Lanthanum, foil, 50x50mm, thickness 0.1mm, as rolled, 99%
La
GF54212914
Lanthanum, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
La