EMAIL THIS PAGE TO A FRIEND
Scientific Reports
Dzip3 regulates developmental genes in mouse embryonic stem cells by reorganizing 3D chromatin conformation.
PMID 26568260
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
80424
Di(N-succinimidyl) glutarate, ≥97.0% (CHN)
C13H14N2O8
EDS
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
C10H16N2O8
E6758
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
C10H16N2O8
431788
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
C10H16N2O8
E9884
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
C10H16N2O8
ED
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
C10H16N2O8
E8008
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
C10H16N2O8
F8775
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
CH2O
252549
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
CH2O
47608
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
CH2O
33220
Formaldehyde solution, puriss. p.a., ACS reagent, ≥36.5%
CH2O
15512
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
CH2O
47083-U
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
CH2O
G3126
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
C5H10N2O3
G5792
L-Glutamine, PharmaGrade, Ajinomoto, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C5H10N2O3
G8540
L-Glutamine, meets USP testing specifications, cell culture tested, 99.0-101.0%, from non-animal source
C5H10N2O3
G6392
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
C5H10N2O3
49419
L-Glutamine, BioUltra, ≥99.5% (NT)
C5H10N2O3
W368401
L-Glutamine
C5H10N2O3
T0397
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
C15H10I4NNaO4 · 5H2O
T2501
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
C15H10I4NNaO4 · 5H2O
449172
Magnesium chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
Cl2Mg
449164
Magnesium chloride, anhydrous, beads, −10 mesh, 99.9% trace metals basis
Cl2Mg
M8266
Magnesium chloride, anhydrous, ≥98%
Cl2Mg
M4880
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Cl2Mg
208337
Magnesium chloride, powder, <200 μm
Cl2Mg
M1028
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Cl2Mg
63069
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Cl2Mg
M8787
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Cl2Mg
63036
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Cl2Mg