EMAIL THIS PAGE TO A FRIEND
Scientific reports
From Soybean residue to advanced supercapacitors.
PMID 26568473
Abstract
Supercapacitor technology is an extremely timely area of research with fierce international competition to develop cost-effective, environmentally friendlier EC electrode materials that have real world application. Herein, nitrogen-doped carbons with large specific surface area, optimized micropore structure and surface chemistry have been prepared by means of an environmentally sound hydrothermal carbonization process using defatted soybean (i.e., Soybean meal), a widely available and cost-effective protein-rich biomass, as precursor followed by a chemical activation step. When tested as supercapacitor electrodes in aqueous electrolytes (i.e. H2SO4 and Li2SO4), they demonstrate excellent capacitive performance and robustness, with high values of specific capacitance in both gravimetric (250-260 and 176 F g(-1) in H2SO4 and Li2SO4 respectively) and volumetric (150-210 and 102 F cm(-3) in H2SO4 and Li2SO4 respectively) units, and remarkable rate capability (>60% capacitance retention at 20 A g(-1) in both media). Interestingly, when Li2SO4 is used, the voltage window is extended up to 1.7 V (in contrast to 1.1 V in H2SO4). Thus, the amount of energy stored is increased by 50% compared to H2SO4 electrolyte, enabling this environmentally sound Li2SO4-based supercapacitor to deliver ~12 Wh kg(-1) at a high power density of ~2 kW kg(-1).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
439126
Acetone, for HPLC, ≥99.9%
C3H6O
W332607
Acetone, ≥99%, meets FCC analytical specifications
C3H6O
W332615
Acetone, natural, ≥97%
C3H6O
01-0520
Acetone, for HPLC
C3H6O
01-0494
Acetone, for residue analysis, JIS 5000
C3H6O
01-0475
Acetone, suitable for determination of dioxins
C3H6O
48358
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
C3H6O
686530
Acetone solution, contains 20.0 % (v/v) acetonitrile, for HPLC
C3H6O
81377
Poly(tetrafluoroethylene), beads
]n
430935
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
]n
430943
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
]n
468096
Poly(tetrafluoroethylene), powder, 35 μm particle size
]n
182478
Poly(tetrafluoroethylene), powder, >40 μm particle size
]n
737992
Poly(tetrafluoroethylene), powder, 200 μm particle size
]n
738670
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
]n
P5958
Potassium hydroxide, BioXtra, ≥85% KOH basis
HKO
306568
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
HKO
757551
Potassium hydroxide, anhydrous, ≥99.97% trace metals basis
HKO
720143
Potassium hydroxide on alumina
HKO
P4494
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
HKO
417661
Potassium hydroxide solution, 45 wt. % in H2O
HKO