PLoS ONE
A Chemical Screening Approach to Identify Novel Key Mediators of Erythroid Enucleation.
PMID 26569102
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
114715
Phenylhydrazine hydrochloride, ≥99%
C6H8N2 · HCl