EMAIL THIS PAGE TO A FRIEND
Sensors (Basel, Switzerland)
A Graphene-Based Biosensing Platform Based on Regulated Release of an Aptameric DNA Biosensor.
PMID 26569239
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0376
Thymine, ≥99%
C5H6N2O2
T0895
Thymine, suitable for cell culture, BioReagent
C5H6N2O2