EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Evidence for Anaerobic Dechlorination of Dechlorane Plus in Sewage Sludge.
PMID 26572321
Abstract
The environmental occurrence of dechlorination moieties from the high production volume flame retardant, Dechlorane Plus (DP), has largely been documented; however, the sources have yet to be well understood. In addition, few laboratory-based studies exist which identify the cause for the occurrence of these chemicals in the environment or humans. Anaerobic dechlorination of the two DP isomers was investigated using a laboratory-simulated wastewater treatment plant (WWTP) environment where anaerobic digestion is used as part of the treatment regime. Known amounts of each isomer were added separately to sewage sludge which provided the electron-donating substrate and at prescribed time points in the incubation, a portion of the media was removed and analyzed for DP and any dechlorination metabolites. After 7 days, monohydrodechlorinated products were observed for both the syn- and anti-DP which were continued throughout the duration of our study (49 days) in an increasing manner giving a calculated formation rate of 0.48 ± 0.09 and 0.79 ± 0.12 pmols/day for syn- and anti-DP, respectively. Furthermore, we observed a second monohydrodechlorinated product only in the anti-DP isomer incubation medium. This strongly suggests that anti-DP is more susceptible to anaerobic degradation than the syn isomer. We also provide compelling evidence to the location of chlorine loss in the dechlorination DP analogues. Finally, the dechlorination DP moieties formed in our study matched the retention times and identification of those observed in surficial sediment located downstream of the WWTP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF07378476
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
GF93740384
Carbon - Vitreous, rod, 100mm, diameter 10mm, glassy carbon
GF05962765
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
GF74135628
Carbon - Vitreous, foil, 10x10mm, thickness 2.0mm, glassy carbon
GF22691426
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
GF96762864
Carbon - Vitreous, foil, 50x50mm, thickness 1.0mm, glassy carbon
GF39873369
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
GF80419035
Carbon - Vitreous, foil, 8x8mm, thickness 0.5mm, glassy carbon
GF61342220
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
GF58042324
Carbon - Vitreous, foam, 275x330mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
GF12408713
Carbon - Vitreous, foam, 275x330mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
GF04031132
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
GF06230922
Carbon - Vitreous, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
GF95357526
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
GF61808889
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
GF09325009
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
GF99431523
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
GF10355958
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
GF89520521
Carbon - Vitreous, foil, 25x25mm, thickness 4.0mm, glassy carbon
GF19690735
Carbon - Vitreous, foil, 50x50mm, thickness 4.0mm, glassy carbon
GF37745275
Carbon - Vitreous, foil, 10mm disks, thickness 0.2mm, glassy carbon
GF43814527
Carbon - Vitreous, foil, 8mm disks, thickness 1.0mm, glassy carbon
GF72189782
Carbon - Vitreous, rod, 100mm, diameter 1.0mm, glassy carbon
GF65427587
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
GF68195403
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon
GF23723423
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
GF91785134
Carbon - Vitreous, rod, 200mm, diameter 10mm, glassy carbon
GF78447707
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
GF45345562
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
GF24425805
Carbon - Vitreous, rod, 200mm, diameter 7.0mm, glassy carbon