EMAIL THIS PAGE TO A FRIEND
Scientific reports
Novel, Synergistic Antifungal Combinations that Target Translation Fidelity.
PMID 26573415
Abstract
There is an unmet need for new antifungal or fungicide treatments, as resistance to existing treatments grows. Combination treatments help to combat resistance. Here we develop a novel, effective target for combination antifungal therapy. Different aminoglycoside antibiotics combined with different sulphate-transport inhibitors produced strong, synergistic growth-inhibition of several fungi. Combinations decreased the respective MICs by ≥8-fold. Synergy was suppressed in yeast mutants resistant to effects of sulphate-mimetics (like chromate or molybdate) on sulphate transport. By different mechanisms, aminoglycosides and inhibition of sulphate transport cause errors in mRNA translation. The mistranslation rate was stimulated up to 10-fold when the agents were used in combination, consistent with this being the mode of synergistic action. A range of undesirable fungi were susceptible to synergistic inhibition by the combinations, including the human pathogens Candida albicans, C. glabrata and Cryptococcus neoformans, the food spoilage organism Zygosaccharomyces bailii and the phytopathogens Rhizoctonia solani and Zymoseptoria tritici. There was some specificity as certain fungi were unaffected. There was no synergy against bacterial or mammalian cells. The results indicate that translation fidelity is a promising new target for combinatorial treatment of undesirable fungi, the combinations requiring substantially decreased doses of active components compared to each agent alone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3514
4,4′-Diisothiocyanatostilbene-2,2′-disulfonic acid disodium salt hydrate, ≥80% (elemental analysis), powder
C16H8N2Na2O6S4 · xH2O
47829
D-(+) Glucose, analytical standard
C6H12O6
G7528
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
C6H12O6
G8270
D-(+)-Glucose, ≥99.5% (GC)
C6H12O6
G5767
D-(+)-Glucose, ACS reagent
C6H12O6
G7021
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
C6H12O6
G6152
D-(+)-Glucose, suitable for mouse embryo, ≥99.5% (GC)
C6H12O6
G5146
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
C6H12O6
49139
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
C6H12O6
47249
D-(+)-Glucose, analytical standard
C6H12O6
D9434
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
C6H12O6
P9297
Paromomycin sulfate salt, ≥98% (TLC)
C23H45N5O14 · xH2SO4
P8692
Paromomycin sulfate salt, plant cell culture tested, BioReagent
C23H45N5O14 · xH2SO4
P5057
Paromomycin sulfate salt, powder, BioReagent, suitable for cell culture, ≥98%
C23H45N5O14 · xH2SO4
792519
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
CHNaO3
S5761
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
CHNaO3
S6297
Sodium bicarbonate, BioXtra, 99.5-100.5%
CHNaO3
S4019
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
CHNaO3
63409
Sodium malonate dibasic, ≥97.0% (NT)
C3H2Na2O4
M4795
Sodium malonate dibasic monohydrate, BioXtra
C3H2Na2O4 · H2O
M1875
Sodium malonate dibasic monohydrate, ≥98% (non-aqueous titration)
C3H2Na2O4 · H2O
O0136
Sodium oxalate, BioXtra
C2Na2O4
71800
Sodium oxalate, puriss. p.a., ACS reagent, ≥99.5% (RT)
C2Na2O4
379735
Sodium oxalate, ≥99.99% trace metals basis
C2Na2O4
223433
Sodium oxalate, ACS reagent, ≥99.5%
C2Na2O4
563188
Sodium thiosulfate, ≥99.99% trace metals basis
Na2O3S2
HT1005
Sodium thiosulfate solution, 2 g/dL in deionized water
Na2O3S2