EMAIL THIS PAGE TO A FRIEND
Scientific reports
Targeted Proteomics Enables Simultaneous Quantification of Folate Receptor Isoforms and Potential Isoform-based Diagnosis in Breast Cancer.
PMID 26573433
Abstract
The distinct roles of protein isoforms in cancer are becoming increasingly evident. FRα and FRβ, two major isoforms of the folate receptor family, generally have different cellular distribution and tissue specificity. However, the presence of FRβ in breast tumors, where FRα is normally expressed, complicates this situation. Prior to applying any FR isoform-based diagnosis and therapeutics, it is essential to monitor the expression profile of FR isoforms in a more accurate manner. An LC-MS/MS-based targeted proteomics assay was developed and validated in this study because of the lack of suitable methodology for the simultaneous and specific measurement of highly homologous isoforms occurring at low concentrations. FRα and FRβ monitoring was achieved by measuring their surrogate isoform-specific peptides. Five human breast cell lines, isolated macrophages and 60 matched pairs of breast tissue samples were subjected to the analysis. The results indicated that FRβ was overexpressed in tumor-associated macrophages (TAMs) but not epithelial cells, in addition to an enhanced level of FRα in breast cancer cells and tissue samples. Moreover, the levels of the FR isoforms were evaluated according to the histology, histopathological features and molecular subtypes of breast cancer. Several positive associations with PR/ER and HER2 status and metastasis were revealed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2886
3-Indoleacetic acid, plant cell culture tested, crystalline
C10H9NO2
I3750
3-Indoleacetic acid, 98%
C10H9NO2
271004
Acetonitrile, anhydrous, 99.8%
C2H3N
675415
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
C2H3N
437557
Acetonitrile, ≥99.5%, ACS reagent
C2H3N
270717
Acetonitrile, HPLC grade, ≥99.93%
C2H3N
L010000-HH
Acetonitrile, for DNA synthesis
C2H3N
01-0650
Acetonitrile, ≥99.8%, for HPLC
C2H3N
01-0623
Acetonitrile, for residue analysis, suitable for 5000 per JIS
C2H3N
01-0645
Acetonitrile, for chromatography
C2H3N
733466
Acetonitrile, electronic grade, 99.999% trace metals basis
C2H3N
L110001
Acetonitrile
C2H3N
L011800
Acetonitrile
C2H3N
639133
Acetonitrile solution, contains 0.1 % (v/v) ammonium hydroxide
C2H3N
576956
Acetonitrile solution, contains 0.1 % (v/v) formic acid, for HPLC
C2H3N
574732
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, for HPLC
C2H3N
574724
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
C2H3N
679186
Acetonitrile solution, contains 5 % (v/v) water, 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, for HPLC
C2H3N
685461
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
C2H3N
703664
Acetonitrile solution, NMR reference standard, 0.23 wt. % in D2O (99.9 atom % D), water 0.05 wt. %, NMR tube size 6.5 mm × 8 in.
C2H3N
D8284
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
C20H10N2Na2O4 · xH2O
43815
DL-Dithiothreitol, BioUltra, for molecular biology, ≥99.5% (RT)
C4H10O2S2
43819
DL-Dithiothreitol, ≥99.0% (RT)
C4H10O2S2
D0632
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
C4H10O2S2
D5545
DL-Dithiothreitol, BioXtra, ≥99.0% (titration)
C4H10O2S2
D9779
DL-Dithiothreitol, for molecular biology, ≥98% (HPLC), ≥99% (titration)
C4H10O2S2
D9163
DL-Dithiothreitol, for electrophoresis, ≥99%
C4H10O2S2
43816
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
C4H10O2S2
646563
DL-Dithiothreitol solution, 1 M in H2O
C4H10O2S2
27002
Formic acid, purum, ≥85%
CH2O2