EMAIL THIS PAGE TO A FRIEND
BMC Infectious Diseases
High prevalence of oxacillinases in clinical multidrug-resistant Acinetobacteri??A?baumannii isolates from the Tshwane region, South Africa - an update.
PMID 26573617
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
471364
1-Dodecanethiol, ≥98%
C12H26S