Oncology reports
Downregulation of Bmi-1 is associated with suppressed tumorigenesis and induced apoptosis in CD44⁺ nasopharyngeal carcinoma cancer stem-like cells.
PMID 26573774
Bmi-1 (B-cell-specific Moloney murine leukemia virus insertion site 1) is a member of the Polycomb group gene (PcG) family, which is involved in the proliferation, migration and tumorigenesis of several types of cancer stem cells (CSCs). However, its precise role and mechanism in CD44+ nasopharyngeal carcinoma (NPC) cancer stem-like cells (CSC-LCs) remain poorly understood. In our previous study, we successfully silenced Bmi-1 by short hairpin RNA (shRNA) in CD44+ NPC CSC-LCs and obtained stable Bmi-1 knockdown (KD) cell lines. In the present study, we tested the cell proliferation by CCK-8 assay and apoptosis by flow cytometry. Scratch wound healing assay, together with Transwell migration and invasion assays were used to measure the migration and invasion capacity. We further evaluated the tumorigenicity of CD44+ NPC CSC-LCs transfected with Bmi-1 shRNA in vivo. Based on our results, knockdown of Bmi-1 by shRNA resulted in the inhibition of tumor proliferation, migration and invasion in vitro, followed by cell apoptosis. In addition, our results preliminarily demonstrated that inhibition of Bmi-1 expression by shRNA increased tumor apoptosis through the p16INK4a-p14ARF-p53 pathway. Bmi-1 silencing in CD44+ NPC CSC-LCs also resulted in the failure to develop tumors in vivo. These results provide important insights into the role of Bmi-1 in the occurrence and development of NPC. Based on our findings, regulation of Bmi-1 in CD44+ NPC CSC-LCs may provide a potential molecular target for the therapy of NPC, and targeted silencing of Bmi-1 by shRNA may have clinical future implications in NPC therapy.