EMAIL THIS PAGE TO A FRIEND
Journal of Neuropathology and Experimental Neurology
A New Mouse Model of Limb-Girdle Muscular Dystrophy Type 2I Homozygous for the Common L276I Mutation Mimicking the Mild Phenotype in Humans.
PMID 26574668