EMAIL THIS PAGE TO A FRIEND
Journal of nanobiotechnology
Near IR fluorescent conjugated poly(ethylene glycol)bisphosphonate nanoparticles for in vivo bone targeting in a young mouse model.
PMID 26577112
Abstract
Bisphosphonate (BP) compounds are widely used in the treatment of bone disorders. This group of drugs with a high affinity to Ca(+2) ions is rapidly attracted to bone mineral, especially in areas of high resorption. We have engineered unique biodegradable BP nanoparticles (NPs) by dispersion co-polymerization of the monomers methacrylate-PEG-BP) and (3-Aminopropyl)mathacrylamide) with the crosslinker monomer tetra ethylene glycol diacrylate. These NPs possess a dual functionality: (1) covalent attachment of a dye (e.g. near IR dye) or a drug to the nanoparticles through the primary amine groups on the surface of the NPs; (2) chelation to the bone mineral hydroxyapatite through the BP on the surface of the NPs. This study describes the uptake of the unique near IR fluorescent Cy 7-conjugated BP NPs in bone of a young mouse model. Blood half-life studies revealed a relatively long half-life (approximately 5 h) due to a high concentration of PEG in the BP NPs as well as a relatively long whole body clearance (approximately 2 weeks). Body distribution studies showed a specific uptake of the BP NPs in bone. These unique engineered BP NPs are planned to be utilized in future work for diagnostic and drug delivery systems that are targeted to bone disorders.