EMAIL THIS PAGE TO A FRIEND
Virology Journal
Actinobacillus pleuropneumoniae induces SJPL cell cycle arrest in G2/M-phase and inhibits porcine reproductive and respiratory syndrome virus replication.
PMID 26577697
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9568
3,3′-Diindolylmethane, ≥98% (HPLC)
C17H14N2