World journal of surgical oncology
Evaluation of RNA-binding motif protein 3 expression in urothelial carcinoma of the bladder: an immunohistochemical study.
PMID 26577765
RNA-binding motif protein 3 (RBM3), involved in cell survival, has paradoxically been linked to both oncogenesis as well as an increased survival in several cancers, including urothelial carcinoma (UCA). The putative prognostic role of RBM3 was studied using cystectomy specimens with 152 invasive UCA with 35 matched metastases, 65 carcinomas in situ (CIS), 22 high-grade papillary UCAs (PAP), and 112 benign urothelium cases. The H-score (HS, staining intensity × % of positive cells) was used for RBM3 immunoexpression. CIS showed the highest HS (mean = 140) followed by benign urothelium (mean = 97). Metastases showed higher HS than primary invasive UCA (P ≤ 0.0001), and high HS was associated with a lower pT stage (P ≤ 0.0001) and a trend toward the absence of lymphovascular invasion (LVI, P = 0.09), but not pN stage (P = 0.35) and surgical margin status (P = 0.81). Univariate analysis (UVA) of disease recurrence only showed an association between pN stage and LVI (P = 0.005 and 0.03, respectively). On UVA of mortality, pT stage was strongly associated with death (P = 0.01) while pN stage, LVI, surgical margin status, and HS were not. Multivariate analysis confirmed the lack of HS association with recurrence (P = 0.08) and death (P = 0.32). Stronger RBM3 immunoexpression correlated with lower stage tumors and a diminished risk for LVI. However, RBM3 does not seem to carry a prognostic significance for clinical outcome (recurrence and mortality). The exact prognostic role of RBM3 in UCA is yet to be determined.