EMAIL THIS PAGE TO A FRIEND
Scientific Reports
Enhancing the methanol tolerance of platinum nanoparticles for the cathode reaction of direct methanol fuel cells through a geometric design.
PMID 26578100