EMAIL THIS PAGE TO A FRIEND
Nature
Therapeutic antibodies reveal Notch control of transdifferentiation in the adult lung.
PMID 26580007
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A79604
4-Aminosalicylic acid, 99%
C7H7NO3
B4429
BIS-TRIS, BioPerformance Certified, cell culture tested, suitable for insect cell culture, ≥98.0%
C8H19NO5
B9754
BIS-TRIS, ≥98.0% (titration)
C8H19NO5
RES1161B-A7
BIS-TRIS, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture
C8H19NO5
PHG0004
BIS-TRIS, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H19NO5
B7535
BIS-TRIS, BioXtra, ≥98.0% (titration)
C8H19NO5
14879
BIS-TRIS, BioUltra, ≥99.0% (NT)
C8H19NO5
C5670
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
CaCl2
429759
Calcium chloride, anhydrous, beads, −10 mesh, ≥99.99% trace metals basis
CaCl2
499609
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
CaCl2
C8106
Calcium chloride, meets USP testing specifications
CaCl2 · 2H2O
449709
Calcium chloride, anhydrous, beads, −10 mesh, ≥99.9% trace metals basis
CaCl2
21115
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
CaCl2
2858
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
C2H6O
48075
Ethanol solution, certified reference material, 2000 μg/mL in methanol
C2H6O
E2385
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
C2H6O
792780
Ethyl alcohol, Pure, 200 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
C2H6O
792799
Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
C2H6O
459836
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
C2H6O
792802
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required
C2H6O
792829
Ethyl alcohol, Pure, 140 proof, meets water USP testing specifications, Excise Tax-free, Permit for use required
C2H6O
792810
Ethyl alcohol, Pure, 140 proof, Excise Tax-free, Permit for use required
C2H6O
493538
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
C2H6O
493511
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
C2H6O
F8775
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
CH2O
F8923
Formaldehyde solution, meets USP testing specifications, contains 9.0-15% methanol as stabilizer
CH2O
252549
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
CH2O
F15587
Formaldehyde solution, 37 wt. % in H2O
CH2O
47608
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
CH2O
33220
Formaldehyde solution, puriss. p.a., ACS reagent, ≥36.5%
CH2O