EMAIL THIS PAGE TO A FRIEND
Asian-Australasian Journal of Animal Sciences
Effect of Suckling Systems on Serum Oxytocin and Cortisol Concentrations and Behavior to a Novel Object in Beef Calves.
PMID 26580289