EMAIL THIS PAGE TO A FRIEND
Scientific reports
Cellular miR-130b inhibits replication of porcine reproductive and respiratory syndrome virus in vitro and in vivo.
PMID 26581169
Abstract
MicroRNAs (miRNAs) can impact viral infections by binding to sequences with partial complementarity on viral RNA transcripts, usually resulting in the repression of virus replication. In the present study, we identified a potential binding site for miR-130 in the 5' untranslated region (bps 155-162) of the porcine reproductive and respiratory syndrome virus (PRRSV) genome. We found that the delivery of multiple miR-130 family mimics, especially miR-130b, resulted in inhibition of PRRSV replication in vitro. miR-130 was effective in inhibiting the replication of multiple type 2 PRRSV strains, but not against vSHE, a classical type 1 strain. miR-130 over-expression did not induce IFN-α or TNF-α expression in either uninfected or PRRSV-infected porcine alveolar macrophages. Results from luciferase reporter assays indicated that miR-130 directly targeted the PRRSV 5' UTR. Intranasal inoculation of piglets with miR-130b exhibited antiviral activity in vivo and partially protected piglets from an otherwise lethal challenge with HP-PRRSV strain vJX143. Overall, these results demonstrate the importance of the miR-130 family in modulating PRRSV replication and also provide a scientific basis for using cellular miRNAs in anti-PRRSV therapies.