EMAIL THIS PAGE TO A FRIEND
BMC Research Notes
Effects of oxygen on the antigenic landscape of prostate cancer cells.
PMID 26581192
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
226947
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
C32H58N2O7S · xH2O
271004
Acetonitrile, anhydrous, 99.8%
C2H3N
675415
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
C2H3N
437557
Acetonitrile, ≥99.5%, ACS reagent
C2H3N
270717
Acetonitrile, HPLC grade, ≥99.93%
C2H3N
L010000-HH
Acetonitrile, for DNA synthesis
C2H3N
01-0650
Acetonitrile, ≥99.8%, for HPLC
C2H3N
01-0623
Acetonitrile, for residue analysis, JIS 5000
C2H3N
01-0645
Acetonitrile, for chromatography
C2H3N
733466
Acetonitrile, electronic grade, 99.999% trace metals basis
C2H3N
L110001
Acetonitrile
C2H3N
L011800
Acetonitrile
C2H3N
639133
Acetonitrile solution, contains 0.1 % (v/v) ammonium hydroxide
C2H3N
576956
Acetonitrile solution, contains 0.1 % (v/v) formic acid, for HPLC
C2H3N
574732
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, for HPLC
C2H3N
574724
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
C2H3N
679186
Acetonitrile solution, contains 5 % (v/v) water, 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, for HPLC
C2H3N
685461
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
C2H3N
703664
Acetonitrile solution, NMR reference standard, 0.23 wt. % in D2O (99.9 atom % D), water 0.05 wt. %, NMR tube size 6.5 mm × 8 in.
C2H3N
43816
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
C4H10O2S2
646563
DL-Dithiothreitol solution, 1 M in H2O
C4H10O2S2
T7875
Thiourea, ReagentPlus®, ≥99.0%
CH4N2S
T8656
Thiourea, ACS reagent, ≥99.0%
CH4N2S
88810
Thiourea, puriss. p.a., ACS reagent, ≥99.0%
CH4N2S
T6508
Trifluoroacetic acid, ReagentPlus®, 99%
C2HF3O2
30-4310
Trifluoroacetic acid, SAJ special grade, ≥99.0%
C2HF3O2
91709
Trifluoroacetic acid solution, BioReagent, suitable for protein sequencing, ~25% in H2O
C2HO2F3
U0631
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
CH4N2O
U5378
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
CH4N2O
U4883
Urea, 8 M (after reconstitution with 16 mL high purity water)
CH4N2O