EMAIL THIS PAGE TO A FRIEND
Scientific reports
Robust Photocatalytic H2O2 Production by Octahedral Cd3(C3N3S3)2 Coordination Polymer under Visible Light.
PMID 26582652
Abstract
Herein, we reported a octahedral Cd3(C3N3S3)2 coordination polymer as a new noble metal-free photocatalyst for robust photocatalytic H2O2 production from methanol/water solution. The coordination polymer can give an unprecedented H2O2 yield of ca. 110.0 mmol • L(-1) • g(-1) at pH = 2.8 under visible light illumination. The characterization results clearly revealed that the photocatalytic H2O2 production proceeds by a pathway of two-electron reduction of O2 on the catalyst surface. This work showed the potential perspective of Mx(C3N3S3)y (M = transitional metals) coordination polymers as a series of new materials for solar energy storage and conversion.