EMAIL THIS PAGE TO A FRIEND
Plant physiology and biochemistry : PPB
Enhanced nodulation of peanut when co-inoculated with fungal endophyte Phomopsis liquidambari and bradyrhizobium.
PMID 26584395
Abstract
In peanut continuous cropping soil, the application of fungal endophyte Phomopsis liquidambari B3 showed peanut pod yield promotion and root nodule number increase. P.xa0liquidambari improved soil environment by degrading allelochemicals and thus promoted peanut pod yield. Furthermore, peanut yield promotion is in part due to the root nodule increase since nodular nitrogen fixation provides the largest source of nitrogen for peanut. However, it is unknown whether this nodule number increase is induced by fungal endophyte. We therefore conducted several pot experiments using vermiculite to investigate the effects of P.xa0liquidambari on peanut-bradyrhizobium nodulation. Our results showed that P.xa0liquidambari co-inoculated with bradyrhizobium increased root nodule number and shoot accumulated nitrogen by 28.25% and 29.71%, respectively. Nodulation dynamics analysis showed that P.xa0liquidambari accelerated nodule initiation and subsequent nodule development. Meanwhile, P.xa0liquidambari was able to colonize the peanut root as an endophyte. The dynamics of P.xa0liquidambari and bradyrhizobial root colonization analysis showed that P.xa0liquidambari inoculation significantly increased the rate of bradyrhizobial colonization. Furthermore, P.xa0liquidambari inoculation significantly increased flavonoids synthesis-related enzymes activities, two common types of flavonoid (luteolin and quercetin-peanut rhizobial nod gene inducer) secretion and lateral root (peanut rhizobial infection site) formation, indicating that P.xa0liquidambari altered the peanut nodulation-related physiological and metabolic activities. These obtained results confirmed the direct contribution of P.xa0liquidambari in enhancing peanut-bradyrhizobium interaction, nodulation and yield.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6013
Cholesteryl hemisuccinate tris salt, anionic detergent
C31H50O4 · C4H11NO3