EMAIL THIS PAGE TO A FRIEND
Scientific Reports
High-Performance Flexible All-Solid-State Supercapacitor from Large Free-Standing Graphene-PEDOT/PSS Films.
PMID 26586106
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
214906
Hypophosphorous acid solution, 50 wt. % in H2O
H3O2P