EMAIL THIS PAGE TO A FRIEND
Scientific Reports
Changes in poly(A) tail length dynamics from the loss of the circadian deadenylase Nocturnin.
PMID 26586468
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282979
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
C7H12O2
A1410
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
C62H86N12O16
A9415
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
C62H86N12O16
A4262
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
C62H86N12O16
01815
Actinomycin D, for fluorescence, ≥90% (HPLC), from Streptomyces sp.
C62H86N12O16
A1895
Aurintricarboxylic acid, practical grade, ≥85% (titration), powder
C22H14O9
A0885
Aurintricarboxylic acid ammonium salt, powder
C22H23N3O9
A36883
Aurintricarboxylic acid ammonium salt, ACS reagent
C22H23N3O9
T0397
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
C15H10I4NNaO4 · 5H2O
T2501
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
C15H10I4NNaO4 · 5H2O
P1639
Patulin, ≥98.0% (HPLC)
C7H6O4
74255
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
C12H25NaO4S
L3771
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
C12H25NaO4S
L6026
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
C12H25NaO4S
436143
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
C12H25NaO4S
L4509
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
C12H25NaO4S
L5750
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
C12H25NaO4S
71725
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
C12H25NaO4S
71729
Sodium dodecyl sulfate, ≥98.0% (GC)
C12H25NaO4S
71717
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
C12H25NaO4S
62862
Sodium dodecyl sulfate, ≥90%
C12H25NaO4S
75746
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
C12H25NaO4S
71736
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
C12H25NaO4S
05030
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
C12H25NaO4S
71687
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
HNaO
757527
Sodium hydroxide, ultra dry, powder, 99.99% trace metals basis
HNaO
72068
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
HNaO
S2567
Sodium hydroxide solution, volumetric, 1.0 M NaOH (1.0N)
HNaO
S2770
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
HNaO
609765
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
HNa16O