EMAIL THIS PAGE TO A FRIEND
Cytotherapy
Intravenous infusion of allogeneic mesenchymal stromal cells in refractory or relapsed aplastic anemia.
PMID 26589752
Abstract
For patients with aplastic anemia (AA) who are refractory to anti-thymocyte globulin (ATG) and cyclosporine, a second course of immunosuppression is successful in only one-fourth to one-third of cases. We conducted a phase 1/2 study to evaluate the addition of two to five weekly intravenous infusions of allogeneic unrelated non-human leukocyte antigen-matched bone marrow-derived mesenchymal stromal cells (MSCs) (median, 2.7xa0× 10(6) cells/kg/infusion; range, 1.3-4.5) to standard rabbit ATG and cyclosporine in nine patients with refractory or relapsed AA. After a median follow-up of 20 months, no infusion-related adverse event was observed, but four deaths occurred as the result of heart failure and bacterial or invasive fungal infections; only two patients achieved partial hematologic responses at 6 months. We failed to demonstrate by fluorescence in situ hybridization or variable number tandem repeat any MSC engraftment in patient marrow 30, 90 or 180 days after infusions. Infusion of allogeneic MSCs in AA is safe but does not improve clinical hematologic response or engraft in recipient bone marrow. This study was registered at clinicaltrials.gov, identifier: NCT01297972.