EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
Nanopolymersomes as potential carriers for rifampicin pulmonary delivery.
PMID 26590894
Abstract
Tuberculosis (TB) has been stated as "the greatest killer worldwide due to a single infectious agent" behind the human immunodeficiency virus. Standard short-term treatment includes the oral administration of a combination of "first-line" drugs. However, poor-patient compliance and adherence to the long-term treatments represent one of the mayor drawbacks of the TB therapy. An alternative to the oral route is the pulmonary delivery of anti-TB drugs for local or systemic administration. Nanotechnology offers an attractive platform to develop novel inhalable/respirable nanocarriers. The present investigation was focused on the encapsulation of rifampicin (RIF) (a "first-line" anti-TB drug) within nanopolymersomes (nanoPS) employing di- and tri-block poly(ethylene glycol) (PEG)-poly(ɛ-caprolactone) (PCL) based copolymers as biomaterials. The derivatives presented a number-average molecular weight between 12.2 KDa and 30.1 KDa and a hydrophobic/hydrophilic balance between 0.56 and 0.99. The nanoPS were able to enhance the apparent RIF aqueous solubility (up to 4.62 mg/mL) where the hydrodynamic diameters of the drug-loaded systems (1% w/v) were ranged between 65.8 nm and 94 nm at day 0 as determined by dynamic light scattering (DLS). Then, RIF-loaded systems demonstrated as excellent colloidal stability in aqueous media over 14 days with a spherical morphology as determined by transmission electron microscopy (TEM). Furthermore, RIF-loaded nano-sized PS promoted drug accumulation in macrophages (RAW 264.7) versus a drug solution representing promising results for a potential TB inhaled therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S3252
Tin(II) 2-ethylhexanoate, 92.5-100.0%
C16H30O4Sn