EMAIL THIS PAGE TO A FRIEND
Talanta
Detection of HIT antibody dependent platelet aggregation using novel surface imprinting approach.
PMID 26592569
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
328634
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%
C5H9NO
494496
1-Methyl-2-pyrrolidinone, biotech. grade, ≥99.7%
C5H9NO
443778
1-Methyl-2-pyrrolidinone, ACS reagent, ≥99.0%
C5H9NO
M79603
1-Methyl-2-pyrrolidinone, ReagentPlus®, 99%
C5H9NO
V3409
1-Vinyl-2-pyrrolidinone, contains sodium hydroxide as inhibitor, ≥99%
C6H9NO
441090
2,2′-Azobis(2-methylpropionitrile), 98%
C8H12N4
755745
2,2′-Azobis(2-methylpropionitrile), recrystallized, 99%
C8H12N4
414565
Divinylbenzene, technical grade, 80%
C10H10
169099
Divinylbenzene, technical grade, 55%
C10H10
F3651
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
C21H11NO5S
46950
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
C21H11NO5S
F4274
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
C21H11NO5S
46951
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
C21H11NO5S
H1758
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
HCl
84429
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
HCl
H9892
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
HCl
2104
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
HCl
W530574
Hydrochloric acid solution, 32 wt. % in H2O, FCC
HCl
295426
Hydrogen chloride, ReagentPlus®, ≥99%
HCl
705551
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
HCl
40104-U Methanolic HCl, 3 M HCl in methanol (3N), for GC derivatization, pkg of 1 × 2 L
HCl
M26305
N-Methylacetamide, ≥99%
C3H7NO
P0115
PAC-1, ≥98% (HPLC)
C23H28N4O2