EMAIL THIS PAGE TO A FRIEND
Talanta
A solid device based on doped hybrid composites for controlling the dosage of the biocide N-(3-aminopropyl)-N-dodecyl-1,3-propanediamine in industrial formulations.
PMID 26592589
Abstract
A colorimetric composite device is proposed to determine the widely used biocide N-(3-aminopropyl)-N-dodecyl-1,3-propanediamine (ADP).This sensing device is based on a film of 1,2-Naphthoquinone-4-sulfonate (NQS) embedded into polydimethylsiloxane-tetraethylortosilicate-SiO2 nanoparticles composite (PDMS-TEOS-SiO2NPs). Semiquantitative analysis can be performed by visual inspection. Digitalized image or diffuse reflectance (DR) measurements can be carried out for quantitative analysis. Satisfactory detection limit (0.018%, w/v) and relative standard deviations <12% were achieved. The proposed device has been applied for the determination of ADP in detergent industrial formulations with recovery values between 80% and 112%. The method has been successfully validated, showing its high potential to control and monitor this compound because the device is easy to prepare and use, robust, portable, stable over time and cost effective. This device allows a green, simple and rapid approach for the analysis of samples without pretreatment and does not require highly trained personnel. These advantages give the proposed kit good prospects for implementation in several industries.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282979
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
C7H12O2
271004
Acetonitrile, anhydrous, 99.8%
C2H3N
675415
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
C2H3N
437557
Acetonitrile, ≥99.5%, ACS reagent
C2H3N
270717
Acetonitrile, HPLC grade, ≥99.93%
C2H3N
L010000-HH
Acetonitrile, for DNA synthesis
C2H3N
01-0650
Acetonitrile, ≥99.8%, for HPLC
C2H3N
01-0623
Acetonitrile, for residue analysis, JIS 5000
C2H3N
01-0645
Acetonitrile, for chromatography
C2H3N
733466
Acetonitrile, electronic grade, 99.999% trace metals basis
C2H3N
L011800
Acetonitrile
C2H3N
576956
Acetonitrile solution, contains 0.1 % (v/v) formic acid, for HPLC
C2H3N
574732
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, for HPLC
C2H3N
574724
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
C2H3N
679186
Acetonitrile solution, contains 5 % (v/v) water, 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, for HPLC
C2H3N
685461
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
C2H3N
703664
Acetonitrile solution, NMR reference standard, 0.23 wt. % in D2O (99.9 atom % D), water 0.05 wt. %, NMR tube size 6.5 mm × 8 in.
C2H3N
H1758
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
HCl
84429
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
HCl
H9892
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
HCl
2104
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
HCl
W530574
Hydrochloric acid solution, 32 wt. % in H2O, FCC
HCl
295426
Hydrogen chloride, ReagentPlus®, ≥99%
HCl
705551
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
HCl
322415
Methanol, anhydrous, 99.8%
CH4O
650609
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
CH4O
270474
Methanol, for HPLC, gradient grade, 99.93%
CH4O
65548
Methanol, for HPLC, gradient grade, ≥99.8% (GC)
CH4O
19-2470
Methanol, for HPLC
CH4O
19-2453
Methanol, for residue analysis, JIS 5000
CH4O