EMAIL THIS PAGE TO A FRIEND
Talanta
Electrochemical immunosensor for ethinylestradiol using diazonium salt grafting onto silver nanoparticles-silica-graphene oxide hybrids.
PMID 26592615
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4906
(+)-Abscisic acid, ≥98% (HPLC)
C15H20O4
E4876
17α-Ethynylestradiol, ≥98%
C20H24O2
A9878
4-Aminobenzoic acid, ReagentPlus®, ≥99%
C7H7NO2
429767
4-Aminobenzoic acid, purified by sublimation, ≥99%
C7H7NO2
100536
4-Aminobenzoic acid, ReagentPlus®, 99%
C7H7NO2
E1253
Estriol, ≥97%
C18H24O3
E1149
Estriol, meets USP testing specifications
C18H24O3
H1758
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
HCl
84429
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
HCl
H9892
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
HCl
2104
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
HCl
W530574
Hydrochloric acid solution, 32 wt. % in H2O, FCC
HCl
H4001
Hydrocortisone, ≥98% (HPLC)
C21H30O5
H0135
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
C21H30O5
H0888
Hydrocortisone, BioReagent, suitable for cell culture
C21H30O5
H3160
Hydrocortisone, meets USP testing specifications
C21H30O5
295426
Hydrogen chloride, ReagentPlus®, ≥99%
HCl
705551
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
HCl
H9003
Hydroquinone, ReagentPlus®, ≥99%
C6H6O2
H3660
Hydroquinone, meets USP testing specifications
C6H6O2
H17902
Hydroquinone, ReagentPlus®, ≥99.5%
C6H6O2
40104-U Methanolic HCl, 3 M HCl in methanol (3N), for GC derivatization, pkg of 1 × 2 L
HCl
P0130
Progesterone, ≥99%
C21H30O2
P6149
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
C21H30O2
P8783
Progesterone, powder, BioReagent, suitable for cell culture
C21H30O2
P3972
Progesterone, meets USP testing specifications
C21H30O2
T1500
Testosterone, ≥98%
C19H28O2
T-037
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material
C19H28O2