EMAIL THIS PAGE TO A FRIEND
Talanta
Selective enrichment and determination of monoamine neurotransmitters by CU(II) immobilized magnetic solid phase extraction coupled with high-performance liquid chromatography-fluorescence detection.
PMID 26592630