EMAIL THIS PAGE TO A FRIEND
Scientific reports
Biophysical Regulation of Chromatin Architecture Instills a Mechanical Memory in Mesenchymal Stem Cells.
PMID 26592929
Abstract
Mechanical cues direct the lineage commitment of mesenchymal stem cells (MSCs). In this study, we identified the operative molecular mechanisms through which dynamic tensile loading (DL) regulates changes in chromatin organization and nuclear mechanics in MSCs. Our data show that, in the absence of exogenous differentiation factors, short term DL elicits a rapid increase in chromatin condensation, mediated by acto-myosin based cellular contractility and the activity of the histone-lysine N-methyltransferase EZH2. The resulting change in chromatin condensation stiffened the MSC nucleus, making it less deformable when stretch was applied to the cell. We also identified stretch induced ATP release and purinergic calcium signaling as a central mediator of this chromatin condensation process. Further, we showed that DL, through differential stabilization of the condensed chromatin state, established a 'mechanical memory' in these cells. That is, increasing strain levels and number of loading events led to a greater degree of chromatin condensation that persisted for longer periods of time after the cessation of loading. These data indicate that, with mechanical perturbation, MSCs develop a mechanical memory encoded in structural changes in the nucleus which may sensitize them to future mechanical loading events and define the trajectory and persistence of their lineage specification.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
16609
1,2-Bis(2-amino-5-methylphenoxy)ethane-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, ≥95% (HPLC)
C36H44N2O18
A4926
1,2-Bis(2-Aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid, 98%
C22H24N2O10
A6535 Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
A1076
BAPTA-AM, ≥95% (HPLC)
C34H40N2O18
C5670
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
CaCl2
429759
Calcium chloride, anhydrous, beads, −10 mesh, ≥99.99% trace metals basis
CaCl2
499609
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
CaCl2
C8106
Calcium chloride, meets USP testing specifications
CaCl2 · 2H2O
449709
Calcium chloride, anhydrous, beads, −10 mesh, ≥99.9% trace metals basis
CaCl2
21115
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
CaCl2
C3662
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
C62H111N11O12
C1832
Cyclosporin A, from Tolypocladium inflatum, BioReagent, for molecular biology, ≥95%
C62H111N11O12
30024
Cyclosporin A, ≥98.5% (HPLC)
C62H111N11O12
E0396
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
C14H24N2O10
E3889
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
C14H24N2O10
E4378
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
C14H24N2O10
I2142
KN-62, ≥95%, powder
C38H35N5O6S2
449172
Magnesium chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
Cl2Mg
449164
Magnesium chloride, anhydrous, beads, −10 mesh, 99.9% trace metals basis
Cl2Mg
M8266
Magnesium chloride, anhydrous, ≥98%
Cl2Mg
M4880
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Cl2Mg
208337
Magnesium chloride, powder, <200 μm
Cl2Mg
M1028
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Cl2Mg
63069
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Cl2Mg
M8787
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Cl2Mg
63036
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Cl2Mg
68475
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Cl2Mg
00457
Magnesium chloride solution, 0.1 M
Cl2Mg
O4876 Oligomycin from Streptomyces diastatochromogenes, ≥90% total oligomycins basis (HPLC)
R2751
Ruthenium Red, Technical grade
H42Cl6N14O2Ru3