EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Comparison of the Efficiency of Viral Transduction and Episomal Transfection in Human Fibroblast Reprogramming.
PMID 26593412
Abstract
Induced pluripotent cells were derived from adult human skin fibroblast by using two methods of reprogramming. Episomal transfection with vectors containing oriP/EBNA-1 sequence for delivery of reprogramming genes Oct4, Sox2, Klf4, L-Myc, and Lin28 proved to be more effective than viral transduction with Sendai virus-based vector: ~200 and 8 colonies per 10(5) cells were found on day 21 of culturing, respectively. Colonies of induced pluripotent cells obtained by these two methods expressed pluripotency marker Tra1-60.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69720
1-Methylxanthine, ≥97.0% (HPLC)
C6H6N4O2