EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Regulation of the Melanocortin-Sensitive Adenylate Cyclase System by N-Acylated Peptide 71-82 of Type 4 Melanocortin Receptor.
PMID 26593415
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T13005
1,2,3,4-Tetrahydroisoquinoline, 95%
C9H11N