EMAIL THIS PAGE TO A FRIEND
Reproductive biology and endocrinology : RB&E
Age-related trends in anti-Mullerian hormone serum level in women with unilateral and bilateral ovarian endometriomas prior to surgery.
PMID 26596960
Abstract
Endometriosis is a well-known cause of infertility, and the anti-Mullerian hormone (AMH) is an accepted biomarker of ovarian reserve and response to artificial reproductive technology procedures. The present study was a prospective analysis of age-dependent AMH serum concentration in women with bilateral and unilateral ovarian endometriomas before therapy onset compared with healthy controls. This prospective cross-sectional study included 384 women aged 18-48 years. AMH serum concentration was assessed between days 3 and 6 of the menstrual cycle in 78 patients with bilateral and 157 patients with unilateral ovarian endometriomas and compared with 149 healthy controls. Ovarian endometriosis was confirmed histopathologically, and data were presented as medians with interquartile range (IQR). Stage III endometriosis was diagnosed in 53.2 %, stage IV in 18.3 %, stage V in 23.4 % and stage VI in 5.4 % of the patients. Patients with bilateral ovarian endometriomas showed the lowest median AMH levels compared with patients suffering from unilateral ovarian endometriosis (0.55; IQR: 0.59 vs. 2.00; IQR: 2.80; p < 0.001) and the control group (0.55; IQR: 0.59 vs. 2.84; IQR: 3.2; p < 0.001). Median AMH concentration values were not significantly different between patients with unilateral ovarian endometriosis and the healthy controls (2.00; IQR: 2.80 vs. 2.84; IQR: 3.2; p = 0.182). A strongly negative correlation between AMH levels and age was confirmed in healthy individuals (R = -0.834; p < 0.001) and women with unilateral ovarian endometriomas (R = -0.774; p < 0.001). Patients with bilateral ovarian endometriosis showed a significantly negative but only moderate correlation between AMH levels and age (R = -0.633; p < 0.001), which was significantly lower than in the healthy controls (R = -0.633 vs. R = -0.834; p = 0.006) but not in the patients with unilateral ovarian endometriosis (R = -0.663 vs. R-0.774; p = 0.093). Based on a multivariate regression analysis, only bilateral localization of ovarian endometrial cysts (p = 0.003) and patient age (p < 0.001), but not left/right localization of unilateral cyst or cyst volume, were negatively associated with AMH serum concentration. According to our data, unilateral ovarian endometriosis had a moderately negative and nonsignificant effect on AMH-based ovarian reserve evaluated prior to surgery, irrespective of age. In contrast, the ovarian reserve was significantly reduced in women with bilateral ovarian endometriomas.