EMAIL THIS PAGE TO A FRIEND
Journal of Neuroinflammation
SDF1-CXCR4 signaling contributes to persistent pain and hypersensitivity via regulating excitability of primary nociceptive neurons: involvement of ERK-dependent Nav1.8 up-regulation.
PMID 26597700