EMAIL THIS PAGE TO A FRIEND
BMC Complementary and Alternative Medicine
Pharmacological basis of the use of the root bark of Zizyphus nummularia Aubrev. (Rhamnaceae) as anti-inflammatory agent.
PMID 26597878