EMAIL THIS PAGE TO A FRIEND
The Journal of Cell Biology
Protocadherins control the modular assembly of neuronal columns in the zebrafish optic tectum.
PMID 26598617
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7629
N-Phenylthiourea, ≥98%
C7H8N2S
222909
N-Phenylthiourea, 97%
C7H8N2S