PLoS ONE
Double-Layer Magnetic Nanoparticle-Embedded Silica Particles for Efficient Bio-Separation.
PMID 26599084
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3648
(3-Aminopropyl)triethoxysilane, ≥98%
C9H23NO3Si
440140
(3-Aminopropyl)triethoxysilane, 99%
C9H23NO3Si
706493
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
C9H23NO3Si
741442
(3-Aminopropyl)triethoxysilane, ≥98.0%
C9H23NO3Si
A7222
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt, ≥96.0% (HPCE), solid
C16H8NNa3O9S3
09341
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt, BioReagent, suitable for fluorescence, ≥96.0% (HPCE)
C16H8NNa3O9S3
H8502
Dopamine hydrochloride
C8H11NO2 · HCl
2858
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
C2H6O
48075
Ethanol solution, certified reference material, 2000 μg/mL in methanol
C2H6O
E2385
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
C2H6O
792780
Ethyl alcohol, Pure, 200 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
C2H6O
792799
Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
C2H6O
459836
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
C2H6O
792802
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required
C2H6O
792829
Ethyl alcohol, Pure, 140 proof, meets water USP testing specifications, Excise Tax-free, Permit for use required
C2H6O
792810
Ethyl alcohol, Pure, 140 proof, Excise Tax-free, Permit for use required
C2H6O
493538
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
C2H6O
493511
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
C2H6O
387649
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
C8H19N
496219
N,N-Diisopropylethylamine, 99.5%, biotech. grade
C8H19N
D125806
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
C8H19N
239690
Succinic anhydride, ≥99% (GC)
C4H4O3