PloS one
Lgr5 Methylation in Cancer Stem Cell Differentiation and Prognosis-Prediction in Colorectal Cancer.
PMID 26599100
Abstract
Leucine-rich-repeat-containing G-protein-coupled receptor 5 (lgr5) is a candidate marker for colorectal cancer stem cells (CSC). In the current study, we investigated the methylation status within thelgr5 promoter and evaluated its relationship with CSC differentiation, prognosis for colorectal cancer, and its clinicopathological features. The methylation status within Lgr5 promoter was detected with a methylation-specific PCR in six colorectal cancer cell lines as well as 169 primary colorectal tumor tissues. Differentiation of CSC was examined with immunofluorescence and immunocytochemistry. Down-regulation of lgr5 was achieved with gene-specific siRNA. The associations between lgr5 methylation and the clinicopathological features as well as survival of patients were analyzed with statistical methods. The lgr5 promoter was methylated to different degrees for the six colorectal cell lines examined, with complete methylation observed in HCT116 cells in which the lgr5 expression was partially recovered following DAC treatment. The stem-cell sphere formation from HCT116 cells was accompanied by increasing methylation within the lgr5 promoter and decreasing expression of lgr5. Knocking down lgr5 by siRNA also led to stem-cell spheres formation. Among primary colorectal tumors, 40% (67/169) were positive for lgr5 methylation, while none of the normal colon tissues were positive for lgr5 methylation. Furthermore, lgr5 methylation significantly associated with higher tumor grade, and negative distant metastasis (p < 0.05), as well as better prognosis (p = 0.001) in patients with colorectal cancer. Our data suggests that lgr5 methylation, through the regulation of lgr5 expression and colorectal CSC differentiation, may constitute a novel prognostic marker for colorectal cancer patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6250
2-Mercaptoethanol, ≥99.0%
C2H6OS
M3148
2-Mercaptoethanol, for molecular biology, for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
C2H6OS
63689
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
C2H6OS
A3656
5-Aza-2′-deoxycytidine, ≥97%
C8H12N4O4
360953
α,α-Dicyanoethyl acetate, 98%
C6H6N2O2
B0126
Bromophenol Blue
C19H10Br4O5S
114391
Bromophenol Blue, ACS reagent
C19H10Br4O5S
G2289
Glycerin, meets USP testing specifications
C3H8O3
G5516
Glycerol, for molecular biology, ≥99.0%
C3H8O3
G6279
Glycerol, BioXtra, ≥99% (GC)
C3H8O3
G9012
Glycerol, ≥99.5%
C3H8O3
G2025
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
C3H8O3
49767
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
C3H8O3
W252506
Glycerol, >99%, FCC, FG
C3H8O3
49781
Glycerol solution, 86-89% (T)
C3H8O3 · xH2O
74255
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
C12H25NaO4S
L3771
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
C12H25NaO4S
L6026
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
C12H25NaO4S
436143
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
C12H25NaO4S
L4509
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
C12H25NaO4S
L5750
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
C12H25NaO4S
71725
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
C12H25NaO4S
71729
Sodium dodecyl sulfate, ≥98.0% (GC)
C12H25NaO4S
71717
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
C12H25NaO4S
62862
Sodium dodecyl sulfate, ≥90%
C12H25NaO4S
75746
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
C12H25NaO4S
71736
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
C12H25NaO4S
05030
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
C12H25NaO4S
S0252
Sulfobromophthalein disodium salt hydrate, used to study hepatocyte transport functions
C20H8Br4Na2O10S2 · xH2O