PloS one
In Vivo Availability of Pro-Resolving Lipid Mediators in Oxazolone Induced Dermal Inflammation in the Mouse.
PMID 26599340
Abstract
The activation and infiltration of polymorphonuclear neutrophils (PMN) are critical key steps in inflammation. PMN-mediated inflammation is limited by anti-inflammatory and pro-resolving mechanisms, including specialized pro-resolving lipid mediators (SPM). We examined the effects of 15-epi-LXA4 on inflammation and the biosynthesis of pro-inflammatory mediators, such as prostaglandins, leukotriene B4 and various hydroxyeicosatetraenoic acids and SPM, in an oxazolone (OXA)-induced hypersensitivity model for dermal inflammation. 15-epi-LXA4 (100 μM, 5 μL subcutaneously injected) significantly (P < 0.05) reduced inflammation in skin, 24 hours after the OXA challenge, as compared to skin treated with vehicle. No significant influence on the biosynthesis of prostaglandins or leukotriene B4 was observed, whereas the level of 15S-hydroxy-eicosatetraenoic acid was significantly (P < 0.05) lower in the skin areas treated with 15-epi-LXA4. In spite of the use of a fully validated analytical procedure, no SPM were detected in the biological samples. To investigate the reason for the lack of analytical signal, we tried to mimic the production of SPM (lipoxins, resolvins, maresin and protectin) by injecting them subcutaneously into the skin of mice and studying the in vivo availability and distribution of the compounds. All analytes showed very little lateral distribution in skin tissue and their levels were markedly decreased (> 95%) 2 hours after injection. However, docosahexaenoic acid derivatives were biologically more stable than SPM derived from arachidonic acid or eicosapentaenoic acid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
862207
4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one, purified by recrystallization
C12H11NO3
439126
Acetone, for HPLC, ≥99.9%
C3H6O
W332615
Acetone, natural, ≥97%
C3H6O
W332607
Acetone, ≥99%, meets FCC analytical specifications
C3H6O
48358
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
C3H6O
686530
Acetone solution, contains 20.0 % (v/v) acetonitrile, for HPLC
C3H6O
270989
Ethyl acetate, anhydrous, 99.8%
C4H8O2
319902
Ethyl acetate, ACS reagent, ≥99.5%
C4H8O2
33211
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
C4H8O2
W241407
Ethyl acetate, ≥99%, FCC, FG
C4H8O2
W241415
Ethyl acetate, natural, ≥99%, FCC, FG
C4H8O2
537446
Ethyl acetate, ReagentPlus®, ≥99.8%
C4H8O2
437549
Ethyl acetate, ACS reagent, ≥99.5%
C4H8O2
270520
Ethyl acetate, HPLC grade, 99.8%
C4H8O2
L092004
Ethyl acetate
C4H8O2
L092003
Ethyl acetate
C4H8O2
L0517
Leukotriene B4, ~100 μg/mL in ethanol, ≥97%
C20H32O4
L0521
Lipoxin A4, ethanol solution
C20H32O5
322415
Methanol, anhydrous, 99.8%
CH4O
650609
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
CH4O
270474
Methanol, for HPLC, gradient grade, 99.93%
CH4O
717975
Methanol, NMR reference standard
CH4O
632546
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, for mass spectrometry, ≥99.5%
CH4O
721964
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
CH4O
721956
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
CH4O
667390
Methanol solution, contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) water, for HPLC
CH4O
650188
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
CH4O